missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Glycine, ≥99%, Molecular biology grade, Calbiochem™,

Catalog No. 3570021KG
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3570021KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3570021KG Supplier MilliporeSigma™ Supplier No. 3570021KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity amino acid suitable for the preparation of biological buffers.

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Color White
Packaging Glass Bottle
Quantity 1 kg
Percent Purity ≥99% (Titration)
Grade Molecular Biology
Physical Form Solid
Chemical Name or Material Glycine