missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Glycine, >99%, Calbiochem™,

Catalog No. 3570500GM
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3570500GM 500 g Glass Bottle
35701KG 1 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 3570500GM Supplier MilliporeSigma™ Supplier No. 3570500GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Color White
Packaging Glass Bottle
Quantity 500 g
Percent Purity >99% (Assay)
Grade Biochemical
Physical Form Crystalline Solid
Chemical Name or Material Glycine