missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycine, 99+%, Cell Culture Reagent

Catalog No. p-7058222
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6240722 100 g
AAJ6240736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6240722 Supplier Thermo Scientific Chemicals Supplier No. J6240722
Only null left

CAS: 56-40-6 | C2H5NO2 | 75.07 g/mol

This buffer solution is used for eluting (dissociating) antibodies in various antibody purification procedures such as affinity purification of antibody using peptide coupled to a column. It is widely used as a buffer for a variety of immunological applications. Glycine is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This buffer solution is used for eluting (dissociating) antibodies in various antibody purification procedures such as affinity purification of antibody using peptide coupled to a column. It is widely used as a buffer for a variety of immunological applications. Glycine is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

Solubility
Soluble in water (100 mg/ml), and acetone (slightly soluble). Insoluble in alcohol, and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature 2 - 8°C

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Melting Point ∽245°C (decomposition)
pH 5.97
Color White
Linear Formula H2NCH2CO2H
Beilstein 635782
Merck Index 14,4491
Solubility Information Soluble in water (100mg/ml),and acetone (slightly soluble). Insoluble in alcohol,and ether.
Density 1.595
Quantity 100 g
Odor Odorless
Formula Weight 75.07
Percent Purity ≥99.5%
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material Glycine
Show More Show Less
EINECSNumber 200-272-2
RTECSNumber MB7600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only