missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycine, 99+%

CAS: 56-40-6 | C2H5NO2 | 75.07 g/mol

$83.56 - $214.14

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA3643530
View Documents Promotion Details
Thermo Scientific Chemicals
03643530
250 g
Each for $83.56
Only null left
 
AA36435A1
View Documents Promotion Details
Thermo Scientific Chemicals
036435A1
1 kg
Each for $214.14
Only null left
 
Description

Description

It is employed to protect the kidneys from the harmful side effects of certain drugs used after organ transplantation as well as the liver from the harmful effects of alcohol. The solution of glycine plays an important role for deposition of alloys and metals. It is likewise useful for patients with acute alcohol hallucinosis. It is applied for treating schizophrenia, stroke, benign prostatic hyperplasia (BPH), and some rare inherited metabolic disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

56-40-6
75.07
DHMQDGOQFOQNFH-UHFFFAOYSA-N
750
2-aminoacetic acid
C2H5NO2
MFCD00008131
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
CHEBI:15428
NCC(O)=O

Specifications

∽245°C (decomposition)
99+%
MFCD00008131
14,4491
Soluble in pyridine,water. Sparingly soluble in ethanol. Insoluble in ether.
NCC(O)=O
2-aminoacetic acid
750
75.07
Powder
56-40-6
C2H5NO2
635782
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
DHMQDGOQFOQNFH-UHFFFAOYSA-N
1.1607
75.07
CHEBI:15428
≥99%
Glycine
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-272-2

RTECSNumber : MB7600000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only