missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycine, 0.2M buffer soln., pH 3.5

Catalog No. aaj60154ap
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60154AP 500 mL
AAJ60154AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60154AP Supplier Thermo Scientific Chemicals Supplier No. J60154AP
Only null left

CAS: 56-40-675.067 g/mol

Glycine, is used as a compound for polyacrylamide gel electrophoresis and Western blots. It is also used widely as a buffer for a variety of immunological applications. It is frequently used in the preparation of TG Buffers (Tris-Glycine; sc-296648) where the buffer is used as a running and/or transfer buffer for polyacrylamide gel electrophoresis and western blotting. It is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine, is used as a compound for polyacrylamide gel electrophoresis and Western blots. It is also used widely as a buffer for a variety of immunological applications. It is frequently used in the preparation of TG Buffers (Tris-Glycine; sc-296648) where the buffer is used as a running and/or transfer buffer for polyacrylamide gel electrophoresis and western blotting. It is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

Solubility
It is soluble in water (100 mg/ml), and acetone (slightly soluble). Insoluble in alcohol, and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong oxidizing agents.

Chemical Identifiers

CAS 56-40-6
Molecular Weight (g/mol) 75.067

Specifications

Concentration 0.2M buffer soln.
pH 3.5
Physical Form Liquid
Quantity 500 mL
Odor Odorless
Recommended Storage Ambient Temperatures

RUO – Research Use Only