missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycine, 0.2M buffer soln., pH 2.5, low endotoxin

Catalog No. p-7057647
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67094AE 100 mL
AAJ67094AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67094AE Supplier Thermo Scientific Chemicals Supplier No. J67094AE
Only null left

CAS: 56-40-675.067 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56-40-6
Molecular Weight (g/mol) 75.067

Specifications

Color Colorless
Concentration 0.2M buffer soln.
pH 2.5
Physical Form Liquid
Quantity 100 mL
Odor Odorless
Recommended Storage Ambient Temperatures

RUO – Research Use Only