missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycinamide Hydrochloride 98.0+%, TCI America™
$125.13 - $365.79
Chemical Identifiers
| CAS | 1668-10-6 |
|---|---|
| Molecular Formula | C2H7ClN2O |
| Molecular Weight (g/mol) | 110.541 |
| MDL Number | MFCD00013008 |
| InChI Key | WKNMKGVLOWGGOU-UHFFFAOYSA-N |
| Synonym | glycinamide hydrochloride, 2-aminoacetamide hydrochloride, glycine amide hydrochloride, h-gly-nh2.hcl, glycinamidehydrochloride, acetamide, 2-amino-, monohydrochloride, glycinamide hcl, glycineamide hcl, glycineamide hydrochloride, glycinamid hydrochloride |
| PubChem CID | 2723639 |
| IUPAC Name | 2-aminoacetamide;hydrochloride |
| SMILES | C(C(=O)N)N.Cl |
Chemical Identifiers
| 1668-10-6 | |
| 110.541 | |
| WKNMKGVLOWGGOU-UHFFFAOYSA-N | |
| 2723639 | |
| C(C(=O)N)N.Cl |
| C2H7ClN2O | |
| MFCD00013008 | |
| glycinamide hydrochloride, 2-aminoacetamide hydrochloride, glycine amide hydrochloride, h-gly-nh2.hcl, glycinamidehydrochloride, acetamide, 2-amino-, monohydrochloride, glycinamide hcl, glycineamide hcl, glycineamide hydrochloride, glycinamid hydrochloride | |
| 2-aminoacetamide;hydrochloride |
Specifications
| 1668-10-6 | |
| White | |
| MFCD00013008 | |
| glycinamide hydrochloride, 2-aminoacetamide hydrochloride, glycine amide hydrochloride, h-gly-nh2.hcl, glycinamidehydrochloride, acetamide, 2-amino-, monohydrochloride, glycinamide hcl, glycineamide hcl, glycineamide hydrochloride, glycinamid hydrochloride | |
| C(C(=O)N)N.Cl | |
| 110.541 | |
| 110.54 | |
| Crystalline Powder |
| 203°C | |
| C2H7ClN2O | |
| 25 g | |
| WKNMKGVLOWGGOU-UHFFFAOYSA-N | |
| 2-aminoacetamide;hydrochloride | |
| 2723639 | |
| ≥98.0% (T) | |
| Glycinamide Hydrochloride |
Safety and Handling
TSCA : No