missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycinamide Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 203°C |
| Color | White |
| Quantity | 25 g |
| Formula Weight | 110.54 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Glycinamide Hydrochloride |
Safety and Handling
| TSCA | No |