missing translation for 'onlineSavingsMsg'
Learn More

Glycinamide hydrochloride, 98%, Thermo Scientific™

Chemical Identifiers

CAS 1668-10-6
Molecular Formula C2H6N2O·HCl
Molecular Weight (g/mol) 110.54
MDL Number MFCD00013008
InChI Key WKNMKGVLOWGGOU-UHFFFAOYSA-N
Synonym glycinamide hydrochloride, 2-aminoacetamide hydrochloride, glycine amide hydrochloride, h-gly-nh2.hcl, glycinamidehydrochloride, acetamide, 2-amino-, monohydrochloride, glycinamide hcl, glycineamide hcl, glycineamide hydrochloride, glycinamid hydrochloride
PubChem CID 2723639
IUPAC Name 2-aminoacetamide;hydrochloride
SMILES C(C(=O)N)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC120060250
SDS View Documents
Thermo Scientific Chemicals
120060250
25g Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC120061000
SDS View Documents
Thermo Scientific Chemicals
120061000
100g Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1668-10-6
110.54
WKNMKGVLOWGGOU-UHFFFAOYSA-N
2723639
C(C(=O)N)N.Cl
C2H6N2O·HCl
MFCD00013008
glycinamide hydrochloride, 2-aminoacetamide hydrochloride, glycine amide hydrochloride, h-gly-nh2.hcl, glycinamidehydrochloride, acetamide, 2-amino-, monohydrochloride, glycinamide hcl, glycineamide hcl, glycineamide hydrochloride, glycinamid hydrochloride
2-aminoacetamide;hydrochloride

Specifications

1668-10-6
Authentic
Plastic bottle
H2NCH2CONH2·HCl
25g
glycinamide hydrochloride, 2-aminoacetamide hydrochloride, glycine amide hydrochloride, h-gly-nh2.hcl, glycinamidehydrochloride, acetamide, 2-amino-, monohydrochloride, glycinamide hcl, glycineamide hcl, glycineamide hydrochloride, glycinamid hydrochloride
C(C(=O)N)N.Cl
110.54
110.54
Glycinamide hydrochloride, 98%
1% max. (105°C, 4 hrs)
97.5% min. (Argentometry)
C2H6N2O·HCl
MFCD00013008
04, 343
WKNMKGVLOWGGOU-UHFFFAOYSA-N
2-aminoacetamide;hydrochloride
2723639
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-789-1

SDS
Documents

Documents

RUO – Research Use Only