missing translation for 'onlineSavingsMsg'
Learn More

Glycinamide hydrochloride, 98%

Catalog No. AAA1112806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1112806 5 g
AAA1112814 25 g
AAA1112822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1112806 Supplier Thermo Scientific Chemicals Supplier No. A1112806
Only null left

CAS: 1668-10-6 | C2H7ClN2O | 110.541 g/mol

Glycinamide hydrochloride is a buffer that is useful in the physiological pH range. And it is also used as a pharmaceutical intermediate, in the organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycinamide hydrochloride is a buffer that is useful in the physiological pH range. And it is also used as a pharmaceutical intermediate, in the organic synthesis.

Solubility
Soluble in water (1100g/L).

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 1668-10-6
Molecular Formula C2H7ClN2O
Molecular Weight (g/mol) 110.541
MDL Number MFCD00013008
InChI Key WKNMKGVLOWGGOU-UHFFFAOYSA-N
Synonym glycinamide hydrochloride, 2-aminoacetamide hydrochloride, glycine amide hydrochloride, h-gly-nh2.hcl, glycinamidehydrochloride, acetamide, 2-amino-, monohydrochloride, glycinamide hcl, glycineamide hcl, glycineamide hydrochloride, glycinamid hydrochloride
PubChem CID 2723639
IUPAC Name 2-aminoacetamide;hydrochloride
SMILES C(C(=O)N)N.Cl

Specifications

Melting Point ∼204°C (decomposition)
Quantity 5 g
Beilstein 3554199
Sensitivity Hygroscopic
Solubility Information Soluble in water (1100g/L).
Formula Weight 110.54
Percent Purity 98%
Chemical Name or Material Glycinamide hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-789-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only