missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl (S)-(+)-3-nitrobenzenesulfonate, 98%

Catalog No. AAH6428914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6428914 25 g
AAH6428906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6428914 Supplier Thermo Scientific Chemicals Supplier No. H6428914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 115314-14-2 | C9H9NO6S | 259.23 g/mol

Glycidyl (S)-(+)-3-nitrobenzenesulfonate is used in the synthesis of enantiopure bisoprolol by reacting with phenols using cesium fluoride as catalyst. It is also involved in the preparation of orally active HIV-1 protease inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycidyl (S)-(+)-3-nitrobenzenesulfonate is used in the synthesis of enantiopure bisoprolol by reacting with phenols using cesium fluoride as catalyst. It is also involved in the preparation of orally active HIV-1 protease inhibitor.

Solubility
Soluble in chloroform, methylene chloride and tetrahydrofuran. Insoluble in hexane.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 115314-14-2
Molecular Formula C9H9NO6S
Molecular Weight (g/mol) 259.23
MDL Number MFCD00064582
InChI Key AIHIHVZYAAMDPM-QMMMGPOBSA-N
Synonym s-+-glycidyl nosylate, s-glycidyl nosylate, s-glycidyl 3-nitrobenzenesulfonate, s-oxiran-2-ylmethyl 3-nitrobenzenesulfonate, s-+-glycidyl 3-nitrobenzenesulfonate, glycidyl 3-nitrobenzenesulfonate, ccris 6394, 2s-+-glycidyl-3-nitrobenzenesulfonate, oxiranylmethyl s-3-nitrobenzenesulfonate, 2s-oxiran-2-yl methyl 3-nitrobenzenesulfonate
PubChem CID 146490
IUPAC Name [(2S)-oxiran-2-yl]methyl 3-nitrobenzenesulfonate
SMILES [O-][N+](=O)C1=CC=CC(=C1)S(=O)(=O)OC[C@@H]1CO1

Specifications

Melting Point 63°C to 66°C
Quantity 25 g
Beilstein 4704457
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform,methylene chloride and tetrahydrofuran. Insoluble in hexane.
Formula Weight 259.23
Percent Purity 98%
Chemical Name or Material Glycidyl (S)-(+)-3-nitrobenzenesulfonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber DB7192010
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only