missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycidyl Propargyl Ether 95.0+%, TCI America™
$55.92 - $225.56
Chemical Identifiers
| CAS | 18180-30-8 |
|---|---|
| Molecular Formula | C6H8O2 |
| Molecular Weight (g/mol) | 112.128 |
| MDL Number | MFCD00099300 |
| InChI Key | SYFZCLMMUNCHNH-UHFFFAOYSA-N |
| Synonym | Glycidyl 2-Propynyl Ether, 2,3-Epoxypropyl Propargyl Ether |
| PubChem CID | 281888 |
| IUPAC Name | 2-(prop-2-ynoxymethyl)oxirane |
| SMILES | C#CCOCC1CO1 |
Chemical Identifiers
| 18180-30-8 | |
| 112.128 | |
| SYFZCLMMUNCHNH-UHFFFAOYSA-N | |
| 281888 | |
| C#CCOCC1CO1 |
| C6H8O2 | |
| MFCD00099300 | |
| Glycidyl 2-Propynyl Ether, 2,3-Epoxypropyl Propargyl Ether | |
| 2-(prop-2-ynoxymethyl)oxirane |
Specifications
| 18180-30-8 | |
| 95°C | |
| MFCD00099300 | |
| Glycidyl 2-Propynyl Ether, 2,3-Epoxypropyl Propargyl Ether | |
| C#CCOCC1CO1 | |
| 112.128 | |
| 112.13 | |
| Liquid |
| Yellow | |
| C6H8O2 | |
| 1 g | |
| SYFZCLMMUNCHNH-UHFFFAOYSA-N | |
| 2-(prop-2-ynoxymethyl)oxirane | |
| 281888 | |
| ≥95.0% (GC) | |
| Glycidyl Propargyl Ether |
Safety and Handling
TSCA : No