accessibility menu, dialog, popup

UserName

CAS RN 18180-30-8

HCOO

CAS RN 18180-30-8

IUPAC Name: 2-[(prop-2-yn-1-yloxy)methyl]oxirane
Synonyms: 2-[(prop-2-yn-1-yloxy)methyl]oxirane
Molecular Weight (g/mol): 112.13
Molecular Formula: C6H8O2
InChi Key: SYFZCLMMUNCHNH-UHFFFAOYNA-N
SMILES: C#CCOCC1CO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results