missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl methacrylate, 97%, stabilized

Catalog No. p-4510438
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC165891000 100 g Glass bottle
AC165895000 500 g Glass bottle
AC165890025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC165891000 Supplier Thermo Scientific Chemicals Supplier No. 165891000

CAS: 106-91-2 | C7H10O3 | 142.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-91-2 , 150-76-5
InChI Key VOZRXNHHFUQHIL-UHFFFAOYSA-N
Synonym glycidyl methacrylate, 2,3-epoxypropyl methacrylate, sy-monomer g, glycidol methacrylate, acriester g, blemmer g, blemmer gma, light ester g, glycidylmethacrylate, 2-methacryloxy methyl oxirane
PubChem CID 7837
IUPAC Name oxiran-2-ylmethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC1CO1

Specifications

Melting Point -41.0°C
Density 1.0700g/mL
Boiling Point 189.0°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C7H10O3
Refractive Index 1.4490 to 1.4510
Quantity 100 g
Specific Gravity 1.07
Solubility Information Solubility in water: insoluble. Other solubilities: miscible in almost all common organic solvents
Molecular Weight (g/mol) 142.15
Stabilizer 50 to 150ppm MEHQ
Viscosity 2.5 mPa.s (20°C)
Formula Weight 142.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Glycidyl methacrylate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
Suspected of causing genetic defects.
May damage fertility.
May cau
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 203-441-9

RUO – Research Use Only