missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl Lauryl Ether 97.0+%, TCI America™

Catalog No. G04481G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G04481G 1 g
G04485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G04481G Supplier TCI America Supplier No. G04481G

Chemical Identifiers

CAS 2461-18-9
Molecular Formula C15H30O2
Molecular Weight (g/mol) 242.403
MDL Number MFCD00022344
InChI Key VMSIYTPWZLSMOH-UHFFFAOYSA-N
Synonym Dodecyl Glycidyl Ether
PubChem CID 17163
IUPAC Name 2-(dodecoxymethyl)oxirane
SMILES CCCCCCCCCCCCOCC1CO1

Specifications

Color Colorless
Boiling Point 324°C
Quantity 1 g
Formula Weight 242.40
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Glycidyl Lauryl Ether
EINECSNumber (2)-2426
TSCA Yes