accessibility menu, dialog, popup

UserName

H3COO

CAS RN 2461-18-9

IUPAC Name: 2-[(dodecyloxy)methyl]oxirane
Synonyms: 2-[(dodecyloxy)methyl]oxirane
Molecular Weight (g/mol): 242.4
Molecular Formula: C15H30O2
InChi Key: VMSIYTPWZLSMOH-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCOCC1CO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results