missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl 4-tert-Butylbenzoate 90.0+%, TCI America™

Catalog No. B138725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B138725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B138725G Supplier TCI America Supplier No. B138725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 59313-58-5
Molecular Formula C14H18O3
Molecular Weight (g/mol) 234.30
MDL Number MFCD00145184
InChI Key NOWVDELPZQQGIG-UHFFFAOYNA-N
Synonym 4-tert-Butylbenzoic Acid Glycidyl Ester
PubChem CID 612972
IUPAC Name (oxiran-2-yl)methyl 4-tert-butylbenzoate
SMILES CC(C)(C)C1=CC=C(C=C1)C(=O)OCC1CO1

Specifications

Color Yellow
Quantity 25 g
Formula Weight 234.30
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Glycidyl 4-tert-Butylbenzoate