accessibility menu, dialog, popup

UserName

H3CH3CCH3OOO

CAS RN 59313-58-5

IUPAC Name: (oxiran-2-yl)methyl 4-tert-butylbenzoate
Synonyms: oxiran-2-ylmethyl 4-tert-butylbenzoate
Molecular Weight (g/mol): 234.3
Molecular Formula: C14H18O3
InChi Key: NOWVDELPZQQGIG-UHFFFAOYNA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C(=O)OCC1CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results