missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl 2-methylphenyl ether, tech. 85%

Catalog No. AAL1013622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1013622 100 g
AAL1013614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1013622 Supplier Thermo Scientific Chemicals Supplier No. L1013622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2210-79-9 | C10H12O2 | 164.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2210-79-9
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00037847
InChI Key KFUSXMDYOPXKKT-UHFFFAOYSA-N
Synonym 2-2-methylphenoxy methyl oxirane, o-cresyl glycidyl ether, glycidyl 2-methylphenyl ether, o-cresol glycidyl ether, glycidyl o-tolyl ether, araldite dy 023, o-kresol-glycidaether, 2-o-tolyloxy methyl oxirane, 1,2-epoxy-3-2-methylphenoxy propane, 1-2-methylphenoxy-2,3-epoxypropane
PubChem CID 16640
IUPAC Name 2-[(2-methylphenoxy)methyl]oxirane
SMILES CC1=CC=CC=C1OCC2CO2

Specifications

Density 1.079
Boiling Point 119°C to 120°C (6 mmHg)
Flash Point 120°C (248°F)
Refractive Index 1.529
Quantity 100 g
UN Number UN3082
Beilstein 4585
Formula Weight 164.21
Percent Purity 85%
Grade Technical
Chemical Name or Material Glycidyl 2-methylphenyl ether
Show More Show Less
Hazard Category H315-H319-H331-H334-H341
Hazard Statement GHS H Statement
H331-H341-H315-H317
Toxic if inhaled.
Suspected of causing genetic defects.
Causes skin irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 218-645-3
RTECSNumber TZ3700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only