accessibility menu, dialog, popup

UserName

CAS RN 2210-79-9

IUPAC Name: 2-[(2-methylphenoxy)methyl]oxirane
Synonyms: cresyl glycidyl ether
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: KFUSXMDYOPXKKT-UHFFFAOYNA-N
SMILES: CC1=CC=CC=C1OCC1CO1
Boiling Point 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)