missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycidamide, ≥95% (qNMR), MilliporeSigma™ Supelco™
Description
Featured Industries: Environmental
Specifications
Specifications
| CAS | 5694-00-8 |
| Linear Formula | C3H5 NO2 |
| MDL Number | MFCD00468053 |
| Synonym | Oxiranecarboxamide |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 87.08 |
| Percent Purity | ≥95% (qNMR) |
| Grade | Analytical Standard |