missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyceryl Monooleate, Spectrum™ Chemical
G1017, 25496-72-4, C21H40O4
Supplier: Spectrum Chemical Mfg Cor G101725LTGL
Description
Spectrum™ Chemical Glyceryl Monooleate is a clear or light yellow oil that is used as an antifoam in juice processing. It has been used as an emulsifier, a moisturizer, and a flavoring agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Specifications
| 25496-72-4 | |
| Glass Bottle | |
| MFCD00067258,MFCD00042735,MFCD00069635 | |
| RZRNAYUHWVFMIP-KTKRTIGZNA-N | |
| 2,3-dihydroxypropyl (9Z)-octadec-9-enoate | |
| Reagent |
| 100% | |
| C21H40O4 | |
| 2.5 L | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)CO | |
| 356.55 |
Chemical Identifiers
| 25496-72-4 | |
| 356.55 | |
| RZRNAYUHWVFMIP-KTKRTIGZNA-N | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)CO |
| C21H40O4 | |
| MFCD00067258,MFCD00042735,MFCD00069635 | |
| 2,3-dihydroxypropyl (9Z)-octadec-9-enoate |