missing translation for 'onlineSavingsMsg'
Learn More

Glyceryl Monooleate, Spectrum™ Chemical

Catalog No. p-7157483
Change view
Click to view available options
Quantity:
500 mL
2.5 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602991 500 mL Amber Glass Bottle
18602992 2.5 L Glass Bottle
18602993 20 L Poly Drum
18602994 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602991 Supplier Spectrum Chemical Mfg Cor Supplier No. G1017500MLGL
Only null left

G1017, 25496-72-4, C21H40O4

Spectrum™ Chemical Glyceryl Monooleate is a clear or light yellow oil that is used as an antifoam in juice processing. It has been used as an emulsifier, a moisturizer, and a flavoring agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 25496-72-4
Molecular Formula C21H40O4
Molecular Weight (g/mol) 356.55
MDL Number MFCD00067258,MFCD00042735,MFCD00069635
InChI Key RZRNAYUHWVFMIP-KTKRTIGZNA-N
IUPAC Name 2,3-dihydroxypropyl (9Z)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)CO

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 500 mL
Grade Reagent