missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyceryl Ascorbate 95.0+%, TCI America™
Supplier: TCI America G045125G
Specifications
| Glyceryl Ascorbate | |
| 159°C | |
| C9H14O8 | |
| 25 g | |
| KQWQJCDIYBPYNT-UHFFFAOYNA-N | |
| 5-(1,2-dihydroxyethyl)-3-(2,3-dihydroxypropoxy)-4-hydroxy-2,5-dihydrofuran-2-one | |
| 133556317 | |
| ≥95.0% (T) |
| 1120360-13-5 | |
| White-Yellow | |
| MFCD28386108 | |
| 2-O-(2,3-Dihydroxypropyl)-L-ascorbic Acid | |
| OCC(O)COC1=C(O)C(OC1=O)C(O)CO | |
| 250.20 | |
| 250.20 | |
| Crystalline Powder |
Chemical Identifiers
| 1120360-13-5 | |
| 250.20 | |
| KQWQJCDIYBPYNT-UHFFFAOYNA-N | |
| 133556317 | |
| OCC(O)COC1=C(O)C(OC1=O)C(O)CO |
| C9H14O8 | |
| MFCD28386108 | |
| 2-O-(2,3-Dihydroxypropyl)-L-ascorbic Acid | |
| 5-(1,2-dihydroxyethyl)-3-(2,3-dihydroxypropoxy)-4-hydroxy-2,5-dihydrofuran-2-one |
Safety and Handling
TSCA : No