missing translation for 'onlineSavingsMsg'
Learn More

Glycerol monoacetate, tech., mixture of isomers, cont. varying amounts of diacetate

Catalog No. AAA1402530
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1402530 250 g
AAA140250B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1402530 Supplier Thermo Scientific Chemicals Supplier No. A1402530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 26446-35-5 | C5H10O4 | 134.13 g/mol

Glycerol monoacetate is used for production of tanning leather and dye. It is also used as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycerol monoacetate is used for production of tanning leather and dye. It is also used as a solvent.

Solubility
Soluble in water, methanol, ethanol, ether (slightly), and insoluble in benzene.

Notes
Hygroscopic. Protect from moisture. Store at room temperature. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 26446-35-5
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.13
MDL Number MFCD00036185
InChI Key KMZHZAAOEWVPSE-UHFFFAOYNA-N
Synonym monoacetin, acetin, 1-monoacetin, glycerol monoacetate, glyceryl monoacetate, acetoglyceride, glyceryl acetate, acetin, 1-mono, monacetin, glycerol 1-monoacetate
PubChem CID 33510
IUPAC Name 2,3-dihydroxypropyl acetate
SMILES CC(=O)OCC(O)CO

Specifications

Density 1.21
Boiling Point 258°C
Flash Point 145°C (293°F)
Refractive Index 1.45
Quantity 250 g
Sensitivity Hygroscopic
Merck Index 14,6242
Solubility Information Soluble in water,methanol,ethanol,ether (slightly),and insoluble in benzene.
Formula Weight 134.13
Grade Technical
Assay cont. varying amounts of diacetate
Chemical Name or Material Glycerol monoacetate, tech., mixture of isomers
Show More Show Less
EINECSNumber 247-704-6
RTECSNumber AK3595000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only