CAS RN 26446-35-5
IUPAC Name:
2,3-dihydroxypropyl acetate
Synonyms:
acetin acetin, 1-mono-
Molecular Weight (g/mol):
134.13
Molecular Formula:
C5H10O4
InChi Key:
KMZHZAAOEWVPSE-UHFFFAOYNA-N
SMILES:
CC(=O)OCC(O)CO
1
–
1
of
1
results
1
Promotions Available
Glycerol monoacetate, tech., mixture of isomers, cont. varying amounts of diacetate
Glycerol monoacetate, tech., mixture of isomers, cont. varying amounts of diacetate
CAS: 26446-35-5 Molecular Formula: C5H10O4 Molecular Weight (g/mol): 134.13 MDL Number: MFCD00036185 InChI Key: KMZHZAAOEWVPSE-UHFFFAOYNA-N Synonym: monoacetin,acetin,1-monoacetin,glycerol monoacetate,glyceryl monoacetate,acetoglyceride,glyceryl acetate,acetin, 1-mono,monacetin,glycerol 1-monoacetate PubChem CID: 33510 IUPAC Name: 2,3-dihydroxypropyl acetate SMILES: CC(=O)OCC(O)CO