missing translation for 'onlineSavingsMsg'
Learn More

Glyburide (Glibenclamide), Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111012311
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012311 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012311 Supplier Merck Emd Millipore Supplier No. PHR12871G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Glyburide (Glibenclamide) is a potent second-generation sulfonylurea widely utilized as an anti-diabetic drug, typically against type 2 diabetes. Its mode of action majorly involves the stimulation of endogenous insulin release from beta cells of pancreas.

Specifications

CAS 10238-21-8
Physical Form Neat
Linear Formula C23H28ClN3O5S
MDL Number MFCD00056625
Synonym Glybenclamide; 5-Chloro-N-[4-(cyclohexylureidosulfonyl)phenethyl]-2-methoxybenzamide; Glyburide; N-p-[2-(5-Chloro-2-methoxybenzamido)ethyl]benzenesulfonyl-N'-cyclohexylurea
Quantity 1 g
Formula Weight 494.00
Grade Certified Reference Material