missing translation for 'onlineSavingsMsg'
Learn More

Glybenzcyclamide, 99%

Catalog No. p-7025787
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2145914 25 g
AAB2145922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2145914 Supplier Thermo Scientific Chemicals Supplier No. B2145914
Only null left

CAS: 10238-21-8 | C23H28ClN3O5S | 494.003 g/mol

Glyburide (Glibenclamide) is a sulfonylurea compound that modulates insulin production. Sulfonylureas bind to ATP-dependent K+ channels in beta cells of the pancreas, depolarizing them and stimulating the release of Ca2+, which in turn stimulates insulin production. Glyburide (Glibenclamide) is an ATP-dependent K+ channel (KIR6, KATP) and CFTR Cl- channel blocker. This compound inhibits recombinant CFTR Cl- channels (IC50 = 20 M).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glyburide (Glibenclamide) is a sulfonylurea compound that modulates insulin production. Sulfonylureas bind to ATP-dependent K+ channels in beta cells of the pancreas, depolarizing them and stimulating the release of Ca2+, which in turn stimulates insulin production. Glyburide (Glibenclamide) is an ATP-dependent K+ channel (KIR6, KATP) and CFTR Cl- channel blocker. This compound inhibits recombinant CFTR Cl- channels (IC50 = 20 μM).

Solubility
Soluble in ethanol (5 mg/mL), DMSO (25 mg/mL), chloroform (1:36), methanol (1:250), and DMF. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 10238-21-8
Molecular Formula C23H28ClN3O5S
Molecular Weight (g/mol) 494.003
MDL Number MFCD00056625
InChI Key ZNNLBTZKUZBEKO-UHFFFAOYSA-N
Synonym glyburide, glibenclamide, glybenclamide, micronase, diabeta, glynase, daonil, euglucon, maninil, semi-daonil
PubChem CID 3488
ChEBI CHEBI:5441
IUPAC Name 5-chloro-N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-2-methoxybenzamide
SMILES COC1=C(C=C(C=C1)Cl)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCCCC3

Specifications

Melting Point 171°C to 174°C
Quantity 25 g
Solubility Information Soluble in ethanol (5mg/mL),DMSO (25mg/mL),chloroform (1:36),methanol (1:250),and DMF. Insoluble in water.
Formula Weight 494.01
Percent Purity 99%
Chemical Name or Material Glybenzcyclamide
EINECSNumber 233-570-6
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only