Learn More
3M™ Gluten Protein Rapid Kit L25GLU, Sold by AquaPhoenix Scientific
For screening for the presence of gluten proteins in clean-in-place (CIP) final rinse water, environmental swab samples, food ingredients and processed food products
Supplier:  3M™ L25GLU
Catalog No. S26914
Only null left
Explore available promotions
Description
Description
  • Results visible at 11 ± 1 min. after application of sample
  • Rapid, qualitative immunochromatographic lateral flow test cartridges designed for detection of allergenic food proteins
  • Designed to detect both processed and unprocessed allergen proteins
  • Test can accommodate environmental swab, CIP rinse water and food samples
Specifications
Specifications
Kit
Allergen
25/Cs.
Gluten Protein Rapid Kit
Screening gluten proteins in CIP final rinse water, environmental swab samples, food ingredients and processed food products
Product Suggestions
Product Suggestions