missing translation for 'onlineSavingsMsg'
Learn More

Glutaronitrile 96.0+%, TCI America™

Supplier:  TCI America G007225G

 View more versions of this product

Catalog No. G007225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Glutaronitrile
Yellow
C5H6N2
25 g
glutaronitrile, 1,3-dicyanopropane, glutarodinitrile, glutaric acid dinitrile, 1,3-trimethylenedinitrile, pyrotartaric acid nitrile, glutaronitrle, unii-01zi68f3cq, trimethylene dicyanide, glutaric dinitrile
C(CC#N)CC#N
94.117
94.12
Liquid
544-13-8
286°C
MFCD00001970
3276
ZTOMUSMDRMJOTH-UHFFFAOYSA-N
pentanedinitrile
10994
≥96.0% (GC)

Chemical Identifiers

544-13-8
94.117
ZTOMUSMDRMJOTH-UHFFFAOYSA-N
10994
C(CC#N)CC#N
C5H6N2
MFCD00001970
glutaronitrile, 1,3-dicyanopropane, glutarodinitrile, glutaric acid dinitrile, 1,3-trimethylenedinitrile, pyrotartaric acid nitrile, glutaronitrle, unii-01zi68f3cq, trimethylene dicyanide, glutaric dinitrile
pentanedinitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : YI3500000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions