missing translation for 'onlineSavingsMsg'
Learn More

Glutaronitrile 96.0+%, TCI America™

Catalog No. p-7130132
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
250 g
Catalog No. Quantity
G007225G 25 g
G0072250G 250 g
2 options

Catalog No. G007225G

Supplier: TCI America G007225G

Only null left

Chemical Identifiers

CAS 544-13-8
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.117
MDL Number MFCD00001970
InChI Key ZTOMUSMDRMJOTH-UHFFFAOYSA-N
Synonym glutaronitrile, 1,3-dicyanopropane, glutarodinitrile, glutaric acid dinitrile, 1,3-trimethylenedinitrile, pyrotartaric acid nitrile, glutaronitrle, unii-01zi68f3cq, trimethylene dicyanide, glutaric dinitrile
PubChem CID 10994
IUPAC Name pentanedinitrile
SMILES C(CC#N)CC#N

Specifications

Color Yellow
Boiling Point 286°C
Quantity 25 g
UN Number 3276
Formula Weight 94.12
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Glutaronitrile
RTECSNumber YI3500000
TSCA Yes