Learn More
Glutarimide, 98%
CAS: 1121-89-7 | C5H7NO2 | 113.12 g/mol
Supplier: Thermo Scientific Chemicals L0096814
Description
Glutarimide acts as an inhibitor of protein synthesis. Further, it is used as a reactant for thionations and biocatalytic asymmetric synthesis of sitagliptin production. It is also employed in the generation of beta-adrenoceptor ligands, enantioselective synthesis of securinega alkaloids and alfa-fluoro-alfa amino amides. In addition to this, it is used in intramolecular amidocyclopropanation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGlutarimide acts as an inhibitor of protein synthesis. Further, it is used as a reactant for thionations and biocatalytic asymmetric synthesis of sitagliptin production. It is also employed in the generation of beta-adrenoceptor ligands, enantioselective synthesis of securinega alkaloids and alfa-fluoro-alfa amino amides. In addition to this, it is used in intramolecular amidocyclopropanation reactions.
Solubility
Soluble in water, hot ethanol and boiling benzene. Insoluble in ether.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Glutarimide | |
| 153°C to 157°C | |
| C5H7NO2 | |
| 25 g | |
| glutarimide, 2,6-piperidinedione, 2,6-diketopiperidine, 2,6-dioxopiperidine, acetylglycinamide impurity b, piperidin-2,6-dione, acmc-2099da, 5-21-09-00557 beilstein handbook reference, 2,6-piperidinedione 9ci | |
| KNCYXPMJDCCGSJ-UHFFFAOYSA-N | |
| piperidine-2,6-dione | |
| 70726 | |
| 113.12 |
| 1121-89-7 | |
| Nearly Odorless | |
| MFCD00006670 | |
| 110052 | |
| Soluble in water,hot ethanol and boiling benzene. Insoluble in ether. | |
| O=C1CCCC(=O)N1 | |
| 113.12 | |
| CHEBI:5435 | |
| 98% |
Chemical Identifiers
| 1121-89-7 | |
| 113.12 | |
| KNCYXPMJDCCGSJ-UHFFFAOYSA-N | |
| 70726 | |
| piperidine-2,6-dione |
| C5H7NO2 | |
| MFCD00006670 | |
| glutarimide, 2,6-piperidinedione, 2,6-diketopiperidine, 2,6-dioxopiperidine, acetylglycinamide impurity b, piperidin-2,6-dione, acmc-2099da, 5-21-09-00557 beilstein handbook reference, 2,6-piperidinedione 9ci | |
| CHEBI:5435 | |
| O=C1CCCC(=O)N1 |
Safety and Handling
EINECSNumber : 214-340-4
RTECSNumber : MA4000000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only