missing translation for 'onlineSavingsMsg'
Learn More

Glutaric Acid 99.0+%, TCI America™

Catalog No. G0069500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0069500G 500 g
G006925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0069500G Supplier TCI America Supplier No. G0069500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-94-1
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.12
MDL Number MFCD00004410
InChI Key JFCQEDHGNNZCLN-UHFFFAOYSA-N
Synonym glutaric acid, 1,5-pentanedioic acid, 1,3-propanedicarboxylic acid, pentandioic acid, glutarate, n-pyrotartaric acid, unii-h849f7n00b, propane-1,3-dicarboxylic acid, c4-c6 dibasic acids, carboxylic acids, c6-18 and c5-15-di
PubChem CID 743
ChEBI CHEBI:17859
IUPAC Name pentanedioic acid
SMILES OC(=O)CCCC(O)=O

Specifications

Melting Point 98°C
Color White
Boiling Point 200°C
Quantity 500 g
Formula Weight 132.12
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Glutaric Acid
EINECSNumber (2)-2560
RTECSNumber MA3740000
TSCA Yes