accessibility menu, dialog, popup

UserName

HOOOHO

CAS RN 110-94-1

IUPAC Name: pentanedioic acid
Synonyms: glutaric acid disodium salt glutaric acid glutarsaeure glutaric acid (substance) n-pyrotartaric acid monosodium salt glutaric acid calcium salt glutaric acid pentanedioc acid ion(1-) glutaric acid glutaric acid, ion(1-)
Molecular Weight (g/mol): 132.12
Molecular Formula: C5H8O4
InChi Key: JFCQEDHGNNZCLN-UHFFFAOYSA-N
SMILES: OC(=O)CCCC(O)=O
Quantity 
  • (2)
  • (3)
  • (4)
Boiling Point 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (6)
Physical Form 
  • (5)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (2)
  • (3)
  • (4)
  • (2)
  • (3)
  • (3)
  • (2)
  • (6)
  • (5)
  • (1)