missing translation for 'onlineSavingsMsg'
Learn More

Glutardialdehyde Solution 25%, For Electron Microscopy, MilliporeSigma™

Catalog No. M1042391000
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1042391000 1 L Glass bottle
M1042392500 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1042391000 Supplier Technidata Supplier No. 1.04239.1000
For electron microscopy

Chemical Identifiers

CAS 111-30-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00007025
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
Synonym glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
PubChem CID 3485
ChEBI CHEBI:64276
IUPAC Name pentanedial
SMILES O=CCCCC=O

Specifications

Physical Form Liquid
Odor Characteristic odor
Packaging Glass bottle
Vapor Pressure 0.27 hPa at 20°C
Color Colorless
Quantity 1 L
Assay Percent Range 25%
UN Number UN 2922
Chemical Name or Material Glutardialdehyde solution
Hazard Category H302 + H332: Harmful if swallowed or if inhaled
H314: Causes severe skin burns and eye damage.
H317: May cause an allergic skin reaction.
H334: May cause allergy or asthma symptoms or breathing difficulties if inhaled.
H335: May cause respiratory irritation.
H400: Very toxic to aquatic life.
DOTInformation DOT Class: 8 (6.1), Packaging Group: II