missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutardialdehyde Solution 25%, For Electron Microscopy, acc. to P.J. Anderson, MilliporeSigma™
$483.64 - $1257.70
Chemical Identifiers
| CAS | 111-30-8 |
|---|---|
| Molecular Formula | C5H8O2 |
| Molecular Weight (g/mol) | 100.12 |
| MDL Number | MFCD00007025 |
| InChI Key | SXRSQZLOMIGNAQ-UHFFFAOYSA-N |
| Synonym | glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde |
| PubChem CID | 3485 |
| ChEBI | CHEBI:64276 |
| IUPAC Name | pentanedial |
| SMILES | O=CCCCC=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
M1121790025
|
Technidata
1121790025 |
25 mL | Glass bottle |
Each for $483.64
|
|
||||
|
M1121790100
|
Technidata
1121790100 |
100 mL | Glass bottle |
Each for $1,257.70
|
|
||||
Description
For electron microscopyChemical Identifiers
| 111-30-8 | |
| 100.12 | |
| SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
| 3485 | |
| pentanedial |
| C5H8O2 | |
| MFCD00007025 | |
| glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde | |
| CHEBI:64276 | |
| O=CCCCC=O |
Specifications
| Liquid | |
| Characteristic odor | |
| 0.27 hPa at 20°C | |
| 25 mL | |
| C5H8O2 | |
| UN 2922 | |
| SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
| pentanedial | |
| 3485 | |
| Glutardialdehyde Solution, Purified and filled under nitrogen |
| 111-30-8 | |
| Glass bottle | |
| Colorless | |
| 25% | |
| MFCD00007025 | |
| glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde | |
| O=CCCCC=O | |
| 100.12 | |
| CHEBI:64276 |
Safety and Handling
H302 + H332: Harmful if swallowed or if inhaled
H314: Causes severe skin burns and eye damage.
H317: May cause an allergic skin reaction.
H334: May cause allergy or asthma symptoms or breathing difficulties if inhaled.
H335: May cause respiratory irritation.
H400: Very toxic to aquatic life.
DOTInformation : DOT Class: 8 (6.1), Packaging Group: II