missing translation for 'onlineSavingsMsg'
Learn More

Glutaraldehyde (ca. 50% in Water, ca. 5.6mol/L), TCI America™

Catalog No. G006825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G006825ML 25 mL
G0068500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G006825ML Supplier TCI America Supplier No. G006825ML

Chemical Identifiers

CAS 111-30-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00007025
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
Synonym glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
PubChem CID 3485
ChEBI CHEBI:64276
IUPAC Name pentanedial
SMILES O=CCCCC=O

Specifications

Physical Form Clear Liquid at 20°C
Color Yellow
pH 3.0 to 6.0
Quantity 25 mL
UN Number 2927
Formula Weight 100.12
Chemical Name or Material Glutaraldehyde (ca. 50% in Water, ca. 5.6mol/L)
EINECSNumber (2)-0509
RTECSNumber MA2450000
TSCA Yes