missing translation for 'onlineSavingsMsg'
Learn More
Learn More
StatLab Glutaraldehyde Buffer 2% pH 7.3
Glutaraldehyde Buffer 2% pH 7.3
$501.04 - $762.43
Specifications
| Physical Form | Solution |
|---|---|
| CAS | 111-30-8 , 124-65-2 |
| Color | Clear Colorless |
| pH | 7.3 |
| Grade | Laboratory |
Alert:
Harmful if swallowed, in contact with skin, or if inhaled
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
22-000-020
|
StatLab
BUG0526 |
100 mL |
Each for $501.04
|
|
|||||
|
22-000-021
|
StatLab
BUG0543 |
16 oz. |
Each for $762.43
|
|
|||||
Description
- Glutaraldehyde Buffer 2% pH 7.3
- A MasterTech Special Stain Kit Component