missing translation for 'onlineSavingsMsg'
Learn More

Glutaraldehyde, 50% Aqueous Solution, MP Biomedicals

Supplier:  MP Biomedicals Inc 0219859580

Catalog No. ICN19859580


Only null left

Description

Description

Specifications

Specifications

Aqueous Solution
111-30-8
100 mL
MFCD00007025
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
pentanedial
3485
100.11
Glutaraldehyde
1.061 g/cm3 (at 20 °C)
C5H8O2
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
O=CCCCC=O
100.12
CHEBI:64276
50%

Chemical Identifiers

111-30-8
100.12
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
3485
pentanedial
C5H8O2
MFCD00007025
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
CHEBI:64276
O=CCCCC=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions