Learn More
Glutaconic aldehyde dianilide hydrochloride, mixture of isomers, 98%
CAS: 1497-49-0 | C17H17ClN2 | 284.79 g/mol
Supplier: Thermo Scientific Chemicals A1839022
Description
Glutaconic aldehyde dianilide hydrochloride is a tricarbocyanine laser dye intermediate. It is used as an intermediate for pharmaceuticals and dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGlutaconic aldehyde dianilide hydrochloride is a tricarbocyanine laser dye intermediate. It is used as an intermediate for pharmaceuticals and dyes.
Solubility
Soluble in methanol. Insoluble in water..
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
Specifications
| Glutaconic aldehyde dianilide hydrochloride | |
| 1497-49-0 | |
| C17H17ClN2 | |
| 100 g | |
| Hygroscopic | |
| Soluble in methanol. Insoluble in water.. | |
| [H+].[Cl-].N(\C=C\C=C\C=NC1=CC=CC=C1)C1=CC=CC=C1 | |
| 284.79 | |
| 284.79 |
| mixture of isomers | |
| ∼124°C to 132°C | |
| MFCD00012630 | |
| 3916924 | |
| hydrogen n-1e,3e-5-phenylimino penta-1,3-dien-1-yl aniline chloride | |
| VUCMMJBDNXZQDJ-ZUJIUJENSA-N | |
| N-[(1E,3E)-5-phenyliminopenta-1,3-dienyl]aniline;hydrochloride | |
| 5702667 | |
| 98% |
Chemical Identifiers
| 1497-49-0 | |
| 284.79 | |
| VUCMMJBDNXZQDJ-ZUJIUJENSA-N | |
| 5702667 | |
| [H+].[Cl-].N(\C=C\C=C\C=NC1=CC=CC=C1)C1=CC=CC=C1 |
| C17H17ClN2 | |
| MFCD00012630 | |
| hydrogen n-1e,3e-5-phenylimino penta-1,3-dien-1-yl aniline chloride | |
| N-[(1E,3E)-5-phenyliminopenta-1,3-dienyl]aniline;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 216-094-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only