missing translation for 'onlineSavingsMsg'
Learn More

Glutaconaldehydedianil Hydrochloride 98.0+%, TCI America™

Catalog No. p-7130188
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G020125G 25 g
G0201250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G020125G missing translation for 'mfr' TCI America missing translation for 'supplierNo' G020125G
Only null left

Chemical Identifiers

CAS 1497-49-0
Molecular Formula C17H17ClN2
Molecular Weight (g/mol) 284.79
MDL Number MFCD00012630
InChI Key VUCMMJBDNXZQDJ-ZUJIUJENSA-N
Synonym hydrogen n-1e,3e-5-phenylimino penta-1,3-dien-1-yl aniline chloride
PubChem CID 5702667
IUPAC Name hydrogen N-[(1E,3E)-5-(phenylimino)penta-1,3-dien-1-yl]aniline chloride
SMILES [H+].[Cl-].N(\C=C\C=C\C=NC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 140°C
Color Purple-Red
Quantity 25 g
Formula Weight 284.79
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Glutaconaldehydedianil Hydrochloride