missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GLUT4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. APP1065
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APP1065 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APP1065 Supplier Invitrogen™ Supplier No. PA11065
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

PA1-1065 detects GLUT4 in rat samples. PA1-1065 has been successfully used in Western blot procedures. By Western blot, this antibody detects a ~46 kDa protein representing GLUT4 in rat heart samples. The PA1-1065 immunogen is a synthetic peptide corresponding to residues E(493) Q E V K P S T E L E Y L G P D E N D(509) of rat GLUT4. The PA1-1065 immunizing peptide (Cat. # PEP-283) is available for use in neutralization and control experiments.

This gene is a member of the solute carrier family 2 (facilitated glucose transporter) family and encodes a protein that functions as an insulin-regulated facilitative glucose transporter. In the absence of insulin, this integral membrane protein is sequestered within the cells of muscle and adipose tissue. Within minutes of insulin stimulation, the protein moves to the cell surface and begins to transport glucose across the cell membrane. Mutations in this gene have been associated with noninsulin-dependent diabetes mellitus (NIDDM).

Specifications

Antigen GLUT4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Slc2a4
Gene Accession No. P19357
Gene Alias glucose transporter 4; Glucose transporter 4 insuline-responsive; Glucose transporter 4, insuline-responsive; glucose transporter GLUT4; glucose transporter type 4; glucose transporter type 4, insulin-responsive; GLUT4; GLUT-4; GT2; insulin responsive glucose transporter; insulin-responsive glucose transporter; insulin-responsive glucose transporter type 4; MGC93607; SLC2A4; solute carrier family 2 member 4; solute carrier family 2 (facilitated glucose transporter), member 4; solute carrier family 2 , member 4; solute carrier family 2 member 4; solute carrier family 2, facilitated glucose transporter member 4
Gene Symbols Slc2a4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues E(493) Q E V K P S T E L E Y L G P D E N D(509) of rat GLUT4.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25139
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less