missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hexanesulfonic Acid Sodium Salt, Monohydrate, 98%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Poly Pail |
| Quantity | 5 kg |
| Molecular Weight (g/mol) | 206.23 |
| Percent Purity | 0.98 |
| Grade | Reagent |
| Physical Form | Crystalline |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.