missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Glucose, high-temperature short-time (HTST)-treated
GREENER_CHOICE

Catalog No. A59774BE
Change view
Click to view available options
Quantity:
100L BPC
200L BPC
20L BPC
50L BPC
5L BPC
Concentration:
25% (w/v)
40% (w/v)
50% (w/v)
Format:
Drum
Sample
Tote
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration Format
A59774BE 200L BPC 40% (w/v) Drum
A59773BA 5L BPC 25% (w/v) Sample
A59773BB 20L BPC 25% (w/v) Tote
A59773BC 50L BPC 25% (w/v) Drum
A59773BD 100L BPC 25% (w/v) Drum
A59773BE 200L BPC 25% (w/v) Drum
A59774BA 5L BPC 40% (w/v) Sample
A59774BB 20L BPC 40% (w/v) Tote
A59774BC 50L BPC 40% (w/v) Drum
A59774BD 100L BPC 40% (w/v) Drum
A59343BA 5L BPC 50% (w/v) Sample
A59343BB 20L BPC 50% (w/v) Tote
A59343BC 50L BPC 50% (w/v) Drum
A59343BD 100L BPC 50% (w/v) Drum
A59343BE 200L BPC 50% (w/v) Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. A59774BE Supplier Gibco™ Supplier No. A59774BE

Gibco™ high-temperature short-time (HTST)-treated products are briefly exposed to temperatures of ≥100°C using in-line pasteurization technology.

Gibco™ high-temperature short-time (HTST)-treated products are briefly exposed to temperatures of ≥100°C using in-line pasteurization technology. HTST is risk mitigation step that may reduce potential viral contaminations from entering upstream manufacturing from raw materials. Products such as glucose and other simple sugars are heated for a controlled period to not alter the treated product’s performance or viability. If HTST is a critical step in your process, outsourcing glucose can help you discover new efficiencies to reclaim time, space, and resources.

Product Features

  • Multiple formulations and concentrations available
  • Available in a range of pack sizes (5 L, 20 L, 50 L, 100 L, and 200 L)
  • Established products with set specifications are made to order
  • Product standardization enables shorter times from inquiry to quote and shipment, compared to custom
  • Readily accessible documentation for accelerated qualification

Benefits

  • Reduces time spent preparing buffers in-house
  • Mitigates space requirement for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Meets workflow needs across stages of development and manufacturing of biologicals

Specifications

Color Clear, colorless solution
Concentration 40% (w/v)
Form Liquid
Product Type Glucose
Sterility Sterile-filtered
Endotoxin Level ≤1.00
Format Drum
Formulation Glucose, HTST-treated, 40% (w/v)
Manufacturing Quality cGMP
Quantity 200L BPC
Tested For Glucose concentration
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.