missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucose-Glutamic Acid, For BOD (Sterilized), Certified, 198.0mg/L BOD ±30.5mg/L, LabChem™
$45.43 - $452.60
Specifications
| Chemical Name or Material | Glucose-Glutamic Acid |
|---|---|
| Color | Colorless |
| CAS Max % | 99.97,0.015,0.015 |
| CAS | 50-99-7,56-86-0 |
| Linear Formula | C6H12O6 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
LC148457
|
LabChem
LC148457 |
125 mL |
Each for $45.43
|
|
|||||
|
LC14845246
|
LabChem
LC14845246 |
24 x 6 mL |
Case of 24 Each for $241.30
|
|
|||||
|
LC14845486
|
LabChem
LC14845486 |
48 x 6 mL |
Each for $452.60
|
|
|||||
|
LC148451
|
LabChem
LC148451 |
500 mL |
Each for $84.47
|
|
|||||
|
LC148452
|
LabChem
LC148452 |
1 L |
Each for $119.57
|
|
|||||
Specifications
| Glucose-Glutamic Acid | |
| 99.97,0.015,0.015 | |
| C6H12O6 | |
| Soluble in water | |
| Certified | |
| Nitrogen oxides; Carbon monoxide; Carbon dioxide |
| Colorless | |
| 50-99-7,56-86-0 | |
| GGA | |
| 180.15 | |
| Passes Test | |
| Liquid |