missing translation for 'onlineSavingsMsg'
Learn More

Glucose-Glutamic Acid, For BOD (Sterilized), Certified, 198.0mg/L BOD ±30.5mg/L, LabChem™

Catalog No. LC148457
Change view
Click to view available options
Quantity:
125 mL
24 x 6 mL
48 x 6 mL
500 mL
1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC148457 125 mL
LC14845246 24 x 6 mL
LC14845486 48 x 6 mL
LC148451 500 mL
LC148452 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC148457 Supplier LabChem Supplier No. LC148457
Only null left

Specifications

Chemical Name or Material Glucose-Glutamic Acid
Color Colorless
Name Note For BOD, Sterilized
CAS Max % 99.97, 0.015, 0.015
CAS 50-99-7, 56-86-0
Linear Formula C6H12O6
Concentration 198.0mg/L BOD ± 30.5mg/L
Synonym GGA
Solubility Information Soluble in water
Formula Weight 180.15
Grade Certified
Identification Passes Test
Packaging Amber Glass
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Quantity 125 mL
Physical Form Liquid
Show More Show Less