missing translation for 'onlineSavingsMsg'
Learn More

Glucosamine hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015121
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015121 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015121 Supplier Merck Emd Millipore Supplier No. PHR1199500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Glucosamine hydrochloride is a drug prescribed for the remediation of degenerative joint diseases in small animals. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 66-84-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C6H13 NO5 · HCl
MDL Number MFCD00135831
Melting Point 190°C to 194°C
Synonym D-(+)-Glucosamine hydrochloride; 2-Amino-2-deoxy-D-glucose hydrochloride; Chitosamine hydrochloride
Type Certified Reference Material
Quantity 500 mg
Formula Weight 215.63
Grade Certified Reference Material
Show More Show Less