missing translation for 'onlineSavingsMsg'
Learn More

Glucosamine Hydrochloride Dietary Supplement, USP, 98-102%, Spectrum™ Chemical

Catalog No. p-7157495
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602932 100 g Amber Glass Bottle
18602933 500 g Amber Glass Bottle
18602934 1 kg Amber Glass Bottle
18602935 5 kg Poly Pail
18602936 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18602932 Supplier Spectrum Chemical Mfg Cor Supplier No. G1094100GM
Only null left

G1094, 66-84-2, C6H13NO5·HCl

Spectrum™ Chemical Glucosamine Hydrochloride, USP Dietary Supplement

Chemical Identifiers

CAS 66-84-2
Molecular Formula C6H14ClNO5
Molecular Weight (g/mol) 215.63
InChI Key CBOJBBMQJBVCMW-UHFFFAOYNA-N
IUPAC Name hydrogen 2-amino-3,4,5,6-tetrahydroxyhexanal chloride
SMILES [H+].[Cl-].NC(C=O)C(O)C(O)C(O)CO

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 1%
Packaging Amber Glass Bottle
Quantity 100 g
Specific Rotation +70.0° to +73.0
Percent Purity 98 to 102%
Grade USP Dietary Supplement