missing translation for 'onlineSavingsMsg'
Learn More

Gluconate Standard, For IC, 1,000 mg/L Gluconate in H2O, MilliporeSigma™ Supelco™

Catalog No. 111004873
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004873 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004873 Supplier Merck Emd Millipore Supplier No. 74549100ML

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST or BAM.

Specifications

Product Line TraceCERT
Concentration 1000 mg/L gluconate aq. soln.
Quantity 100 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC