missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ GloWear™ 8918BK Class E Black Bottom Rain Bibs

Class E Black Bottom Rain Bibs provide breathable, waterproof, comfort in a bib style with black accents to hide dirt.

$105.98 - $105.98

Specifications

Material Polyurethane
Color Yellow
Waist Style Adjustable elastic suspenders
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
View More Specs

Products 8
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-820-901
View Documents
Ergodyne™
25012
Small
Each for $105.98
Only null left
 
19-820-902
View Documents
Ergodyne™
25013
Medium
Each for $105.98
Only null left
 
19-820-903
View Documents
Ergodyne™
25014
Large
Each for $105.98
Only null left
 
19-820-904
View Documents
Ergodyne™
25015
X-Large
Each for $105.98
Only null left
 
19-820-908
View Documents
Ergodyne™
25016
2X-Large
Each for $105.98
Only null left
 
19-820-911
View Documents
Ergodyne™
25017
3X-Large
Each for $105.98
Only null left
 
19-820-916
View Documents
Ergodyne™
25018
4X-Large
Each for $105.98
Only null left
 
19-820-917
View Documents
Ergodyne™
25019
5X-Large
Each for $105.98
Only null left
 
Description

Description

  • Weatherproof 300D oxford outer shell
  • Breathable PU coating
  • 2 in. ANSI-compliant 3M™ Scotchlite™ Reflective
  • Adjustable elastic suspenders
  • Zippered leg openings for easy on/off
  • One right leg cargo pocket
  • Black accents on bottom of leg
Specifications

Specifications

Polyurethane
Yellow
Adjustable elastic suspenders
Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'